accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3HS

CAS RN 25360-10-5

IUPAC Name: 2-methyloctane-2-thiol
Synonyms: tert-nonanethiol
Molecular Weight (g/mol): 160.32
Molecular Formula: C9H20S
InChi Key: MPBLPZLNKKGCGP-UHFFFAOYSA-N
SMILES: CCCCCCC(C)(C)S
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results