accessibility menu, dialog, popup

UserName

Viewing profile as user:
RuPPCPOHHPhPhPhPhPhPhPhPhPh

CAS RN 25360-32-1

IUPAC Name: methanidylidyneoxidanium tris(triphenylphosphane) λ²-ruthenium
Synonyms: carbon monoxide tris(triphenylphosphine) λ²-ruthenium
Molecular Weight (g/mol): 917.97
Molecular Formula: C55H47OP3Ru
InChi Key: MLIYPCQSOXNTLJ-UHFFFAOYSA-N
SMILES: [RuH2].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)