accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNSHN

CAS RN 25369-78-2

IUPAC Name: 5-chloro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-chloro-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 184.64
Molecular Formula: C7H5ClN2S
InChi Key: ZZIHEYOZBRPWMB-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC(=S)NC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)