accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 25372-03-6

IUPAC Name: 4-(1H-imidazol-1-yl)benzonitrile
Synonyms: 4-(imidazol-1-yl)benzonitrile
Molecular Weight (g/mol): 169.19
Molecular Formula: C10H7N3
InChi Key: GYFGZFJGMRRTTP-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)N1C=CN=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results