accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOH

CAS RN 25379-88-8

IUPAC Name: methyl 2-(3,4-dihydroxyphenyl)acetate
Synonyms: methyl 2-(3,4-dihydroxyphenyl)acetate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: UGFILLIGHGZLHE-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC(O)=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results