accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 254-60-4

IUPAC Name: 1,8-naphthyridine
Synonyms: 1,8-naphthyridine 1,8-pyridopyridine napy
Molecular Weight (g/mol): 130.15
Molecular Formula: C8H6N2
InChi Key: FLBAYUMRQUHISI-UHFFFAOYSA-N
SMILES: C1=CC2=CC=CN=C2N=C1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results