accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 254-79-5

IUPAC Name: 1,5-naphthyridine
Synonyms: 1,5-naphthyridine 1,5-pyridopyridine
Molecular Weight (g/mol): 130.15
Molecular Formula: C8H6N2
InChi Key: VMLKTERJLVWEJJ-UHFFFAOYSA-N
SMILES: C1=CC2=NC=CC=C2N=C1
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)