accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 2541-69-7

IUPAC Name: 7-methyltetraphene
Synonyms: benz(A)anthracene-7-methyl
Molecular Weight (g/mol): 242.32
Molecular Formula: C19H14
InChi Key: DIIFUCUPDHMNIV-UHFFFAOYSA-N
SMILES: CC1=C2C=CC3=CC=CC=C3C2=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results