accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOHNOHOOO

CAS RN 254101-10-5

IUPAC Name: 5-(benzyloxy)-3-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(benzyloxy)-3-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 337.37
Molecular Formula: C17H23NO6
InChi Key: FAFJSSKTLCNWRJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)CC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)