accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHCH3

CAS RN 25419-06-1

IUPAC Name: methyl(pentyl)amine
Synonyms: methyl(pentyl)amine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: UOIWOHLIGKIYFE-UHFFFAOYSA-N
SMILES: CCCCCNC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)