accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 254732-51-9

IUPAC Name: 2-chloro-3-(trifluoromethyl)quinoxaline
Synonyms: 2-chloro-3-(trifluoromethyl)quinoxaline
Molecular Weight (g/mol): 232.59
Molecular Formula: C9H4ClF3N2
InChi Key: DSMMAQWRRJQVTQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC2=CC=CC=C2N=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results