accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOOCH2

CAS RN 2549-51-1

IUPAC Name: ethenyl 2-chloroacetate
Synonyms: ethenyl 2-chloroacetate
Molecular Weight (g/mol): 120.53
Molecular Formula: C4H5ClO2
InChi Key: XJELOQYISYPGDX-UHFFFAOYSA-N
SMILES: ClCC(=O)OC=C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)