accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 254966-93-3

IUPAC Name: potassium acetate
Synonyms: potassium acetate|KOAc|AcOK|kaliumazetat|MeCO2K
Molecular Weight (g/mol): 98.14
Molecular Formula: C2H3KO2
InChi Key: SCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES: [K+].CC([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results