accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiOH3COH3COH3C

CAS RN 2550-02-9

IUPAC Name: triethoxy(propyl)silane
Synonyms: triethoxy(propyl)silane
Molecular Weight (g/mol): 206.36
Molecular Formula: C9H22O3Si
InChi Key: NBXZNTLFQLUFES-UHFFFAOYSA-N
SMILES: CCC[Si](OCC)(OCC)OCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results