accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2550-40-5

IUPAC Name: (cyclohexyldisulfanyl)cyclohexane
Synonyms: dicyclohexyl disulfide
Molecular Weight (g/mol): 230.43
Molecular Formula: C12H22S2
InChi Key: ODHAQPXNQDBHSH-UHFFFAOYSA-N
SMILES: C1CCC(CC1)SSC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results