accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25503-91-7

IUPAC Name: 1-acetylpiperidine-4-carbonitrile
Synonyms: 1-acetylpiperidine-4-carbonitrile
Molecular Weight (g/mol): 152.2
Molecular Formula: C8H12N2O
InChi Key: NFDGRMQIOHRQHF-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC(CC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results