accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHOOOHNH2

CAS RN 25548-16-7

IUPAC Name: 3-amino-3-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)carbamoyl]propanoic acid
Synonyms: aminosweet
Molecular Weight (g/mol): 294.31
Molecular Formula: C14H18N2O5
InChi Key: IAOZJIPTCAWIRG-UHFFFAOYNA-N
SMILES: COC(=O)C(CC1=CC=CC=C1)NC(=O)C(N)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results