accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2555-29-5

H3COHOOOCH3

CAS RN 2555-29-5

IUPAC Name: 8-acetyl-7-hydroxy-4-methyl-2H-chromen-2-one
Synonyms: 8-acetyl-7-hydroxy-4-methylchromen-2-one
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: WZOMQVFUPMLOGT-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2OC(=O)C=C(C)C2=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results