accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 2555-49-9

IUPAC Name: ethyl 2-phenoxyacetate
Synonyms: ethyl phenoxyacetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: MGZFVSUXQXCEHM-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)