accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 2556-73-2

IUPAC Name: 1-methylazepan-2-one
Synonyms: 1-methylazepan-2-one
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: ZWXPDGCFMMFNRW-UHFFFAOYSA-N
SMILES: CN1CCCCCC1=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)