accessibility menu, dialog, popup

UserName

Viewing profile as user:
FS

CAS RN 2557-78-0

IUPAC Name: (2-fluorophenyl)sulfanide
Synonyms: (2-fluorophenyl)sulfanide
Molecular Weight (g/mol): 127.16
Molecular Formula: C6H4FS
InChi Key: WJTZZPVVTSDNJJ-UHFFFAOYSA-M
SMILES: FC1=CC=CC=C1[S-]
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)