accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 255724-71-1

IUPAC Name: 5-fluoro-2-iodoaniline
Synonyms: 5-fluoro-2-iodoaniline
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: UCPDOOZBROQHME-UHFFFAOYSA-N
SMILES: NC1=C(I)C=CC(F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results