accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CPCH3CH3H3C(?)

CAS RN 255836-67-0

IUPAC Name: {[1,1'-binaphthalen]-2-yl}di-tert-butyl)phosphane
Synonyms: [1,1'-binaphthalen]-2-yldi-tert-butyl)phosphane
Molecular Weight (g/mol): 398.53
Molecular Formula: C28H31P
InChi Key: QGBQGMHXBSLYLZ-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1)C(C)(C)C
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results