accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PCH3CH3H3CCH3CH3H3C

CAS RN 255837-19-5

IUPAC Name: di-tert-butyl({2'-methyl-[1,1'-biphenyl]-2-yl})phosphane
Synonyms: di-tert-butyl({2'-methyl-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 312.44
Molecular Formula: C21H29P
InChi Key: UJONYAVMBYXBJQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=CC=CC=C1P(C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)