accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN

CAS RN 25594-62-1

IUPAC Name: 1-(quinoxalin-2-yl)ethan-1-one
Synonyms: 1-(quinoxalin-2-yl)ethanone
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: SWGILLQQUULMJB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CN=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results