accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25596-24-1

IUPAC Name: trimethyl(oxo)-λ⁶-sulfanylium bromide
Synonyms: trimethyl(oxo)-λ⁶-sulfanylium bromide
Molecular Weight (g/mol): 173.07
Molecular Formula: C3H9BrOS
InChi Key: KEPJZBFFLDRKSF-UHFFFAOYSA-M
SMILES: [Br-].C[S+](C)(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results