accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 25603-67-2

IUPAC Name: 9-phenyl-9H-fluoren-9-ol
Synonyms: 9-phenylfluoren-9-ol
Molecular Weight (g/mol): 258.32
Molecular Formula: C19H14O
InChi Key: UJPHBDAPVWFPTG-UHFFFAOYSA-N
SMILES: OC1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)