accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOOH2C

CAS RN 25608-12-2

IUPAC Name: potassium prop-2-enoate
Synonyms: potassium polyacrylate
Molecular Weight (g/mol): 110.15
Molecular Formula: C3H3KO2
InChi Key: ZUBIJGNKOJGGCI-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)