accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHF

CAS RN 2561-17-3

IUPAC Name: 1-ethynyl-3-fluorobenzene
Synonyms: 1-ethynyl-3-fluorobenzene
Molecular Weight (g/mol): 120.13
Molecular Formula: C8H5F
InChi Key: PTRUTZFCVFUTMW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C#C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)