accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N(S)

CAS RN 25611-78-3

IUPAC Name: (1S)-1,2-diphenylethan-1-aminium
Synonyms: (1S)-1,2-diphenylethanaminium
Molecular Weight (g/mol): 198.29
Molecular Formula: C14H16N
InChi Key: DTGGNTMERRTPLR-AWEZNQCLSA-O
SMILES: [NH3+][C@@H](CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results