accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25617-97-4

IUPAC Name: gallanylidyneamine
Synonyms: gallium nitride
Molecular Weight (g/mol): 83.73
Molecular Formula: GaN
InChi Key: JMASRVWKEDWRBT-UHFFFAOYSA-N
SMILES: N#[Ga]
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)