accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2562-71-2

IUPAC Name: 4-(1H-1,3-benzodiazol-2-yl)-N,N-dimethylaniline
Synonyms: 4-(1H-1,3-benzodiazol-2-yl)-N,N-dimethylaniline
Molecular Weight (g/mol): 237.31
Molecular Formula: C15H15N3
InChi Key: ZKBBGUJBGLTNEK-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results