accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 25662-28-6

IUPAC Name: methyl cyclopent-1-ene-1-carboxylate
Synonyms: methyl cyclopent-1-ene-1-carboxylate
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: VTYCAXIAUKEGBQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CCCC1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)