accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSH

CAS RN 25697-55-6

IUPAC Name: [3-(trifluoromethyl)phenyl]methanethiol
Synonyms: [3-(trifluoromethyl)phenyl]methanethiol
Molecular Weight (g/mol): 192.2
Molecular Formula: C8H7F3S
InChi Key: CQIQWIMXCPTQPJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CS)=C1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results