accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25710-20-7

IUPAC Name: 2,3-diamino-5-chloropyridin-1-ium
Synonyms: 2,3-diamino-5-chloropyridin-1-ium
Molecular Weight (g/mol): 144.58
Molecular Formula: C5H7ClN3
InChi Key: ZRCMCGQDIYNWDX-UHFFFAOYSA-O
SMILES: NC1=CC(Cl)=C[NH+]=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results