accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2572-25-0

H3COHH3COHH3COHGa

CAS RN 2572-25-0

IUPAC Name: tris(ethanol) gallium
Synonyms: tris(ethyl alcohol) gallium
Molecular Weight (g/mol): 207.93
Molecular Formula: C6H18GaO3
InChi Key: JWISYUBLQXUTOS-UHFFFAOYSA-N
SMILES: [Ga].CCO.CCO.CCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results