accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25726-99-2

IUPAC Name: 3-(dibutylamino)propanenitrile
Synonyms: 3-(dibutylamino)propanenitrile
Molecular Weight (g/mol): 182.31
Molecular Formula: C11H22N2
InChi Key: XNHGXTPCDNEDCH-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)CCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results