accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiOSiOSiOSiOH3COOCH3OOCH3OOOO

CAS RN 257284-60-9

IUPAC Name: 2,4,6,8-tetramethyl-2,4,6,8-tetrakis({3-[(oxiran-2-yl)methoxy]propyl})-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Synonyms: 2,4,6,8-tetramethyl-2,4,6,8-tetrakis[3-(oxiran-2-ylmethoxy)propyl]-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Weight (g/mol): 697.08
Molecular Formula: C28H56O12Si4
InChi Key: BLZDTVROPKIJLZ-UHFFFAOYNA-N
SMILES: C[Si]1(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)