CAS RN 25764-10-7
IUPAC Name:
lanthanioamine
Synonyms:
lanthanioamine
Molecular Weight (g/mol):
152.91
Molecular Formula:
LaN
InChi Key:
QCLQZCOGUCNIOC-UHFFFAOYSA-N
SMILES:
N#[La]
1
–
1
of
1
results
1
Promotions Available
Lanthanum(III) nitride, 99.9% (REO)
Lanthanum(III) nitride, 99.9% (REO)
CAS: 25764-10-7 Molecular Formula: LaN Molecular Weight (g/mol): 152.912 MDL Number: MFCD00049962 InChI Key: QCLQZCOGUCNIOC-UHFFFAOYSA-N Synonym: lanthanum nitride,lanthanum iii nitride,lanthanumylidyneamine PubChem CID: 117627 IUPAC Name: azanylidynelanthanum SMILES: N#[La]