accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25764-11-8

IUPAC Name: neodymioamine
Synonyms: neodymioamine
Molecular Weight (g/mol): 158.25
Molecular Formula: NNd
InChi Key: OVMJQLNJCSIJCH-UHFFFAOYSA-N
SMILES: N#[Nd]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results