accessibility menu, dialog, popup

UserName

Viewing profile as user:
NGd

CAS RN 25764-15-2

IUPAC Name: gadolinioamine
Synonyms: gadolinioamine
Molecular Weight (g/mol): 171.26
Molecular Formula: GdN
InChi Key: FLATXDRVRRDFBZ-UHFFFAOYSA-N
SMILES: N#[Gd]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results