accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25773-40-4

IUPAC Name: 2-methoxy-3-(propan-2-yl)pyrazine
Synonyms: 2-methoxy-3-isopropylpyrazine isopropyl methoxy pyrazine isopropyl-methoxypyrazine
Molecular Weight (g/mol): 152.2
Molecular Formula: C8H12N2O
InChi Key: NTOPKICPEQUPPH-UHFFFAOYSA-N
SMILES: COC1=NC=CN=C1C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results