accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNSOHO

CAS RN 257876-07-6

IUPAC Name: 2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 274.12
Molecular Formula: C10H5Cl2NO2S
InChi Key: ANTJCNIMRNBFMJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC(=N1)C1=C(Cl)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results