accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 257932-29-9

IUPAC Name: 2-hydroxybicyclo[3.2.1]octane-6-carboxylic acid
Synonyms: 2-hydroxybicyclo[3.2.1]octane-6-carboxylic acid
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: FSQSHXLJRFYYMV-UHFFFAOYNA-N
SMILES: OC1CCC2CC1CC2C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results