accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOO

CAS RN 2581-34-2

IUPAC Name: 3-methyl-4-nitrophenol
Synonyms: 3-methyl-4-nitrophenol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: PIIZYNQECPTVEO-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)