accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OH3CCH3OHO

CAS RN 25812-30-0

IUPAC Name: 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
Synonyms: gemfibrozil ausgem nu-pharm brand of gemfibrozil novo gemfibrozil bayvit, gemfibrozilo 1a brand of gemfibrozil pms-gemfibrozil WL-gemfibrozil gemfi 1A pharma GenRX Gemfibrozil
Molecular Weight (g/mol): 250.34
Molecular Formula: C15H22O3
InChi Key: HEMJJKBWTPKOJG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)