accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25830-77-7

IUPAC Name: 2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(2,6-dioxooxan-3-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 259.22
Molecular Formula: C13H9NO5
InChi Key: ICDLEMPZXFCQEB-UHFFFAOYNA-N
SMILES: O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results