accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOONOOOAl

CAS RN 25838-59-9

IUPAC Name: aluminium(3+) trinitrate
Synonyms: aluminium nitrate
Molecular Weight (g/mol): 212.99
Molecular Formula: AlN3O9
InChi Key: JLDSOYXADOWAKB-UHFFFAOYSA-N
SMILES: [Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)