accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2584-71-6

IUPAC Name: (2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: cis-4-hydroxy-D-proline
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: PMMYEEVYMWASQN-QWWZWVQMSA-N
SMILES: O[C@H]1CN[C@H](C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)