accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSOHO

CAS RN 25840-82-8

IUPAC Name: 2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms: 2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 363.48
Molecular Formula: C22H21NO2S
InChi Key: DLMYFMLKORXJPO-UHFFFAOYNA-N
SMILES: NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)